Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEGF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032445.2 | NP_115821.2 | 23 | 4910 | ||
XM_017022670.1 | XP_016878159.1 | 24 | 5491 | |||
XM_017022671.1 | XP_016878160.1 | 24 | 6251 | |||
XM_017022672.1 | XP_016878161.1 | 22 | 5339 | |||
XM_017022674.1 | XP_016878163.1 | 20 | 5594 | |||
XM_017022675.1 | XP_016878164.1 | 20 | 5632 | |||
GenBank mRNA | AB058677.1 | - | 21 | 4703 | ||
AK090663.1 | - | 1 | 1044 |