Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031714.3 | NP_001026884.3 | 4 | 738 | ||
NM_022489.3 | NP_071934.3 | 4 | 738 | |||
NM_032714.2 | NP_116103.1 | 4 | 738 | |||
XM_005268004.4 | XP_005268061.1 | 4 | 825 | |||
XM_005268005.4 | XP_005268062.1 | 4 | 796 | |||
XM_017021595.1 | XP_016877084.1 | 4 | 796 | |||
GenBank mRNA | AK290083.1 | - | 4 | 723 | ||
BC006173.1 | - | 4 | 723 | |||
BI489810.1 | - | 4 | 739 | |||
BQ961756.1 | - | 1 | 24 | |||
BX248757.1 | - | 4 | 707 |