Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEDAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032849.3 | NP_116238.2 | 2 | 642 | ||
GenBank mRNA | AB055407.1 | - | 1 | 28 | ||
AK055635.1 | - | 2 | 626 | |||
BC040165.1 | - | 2 | 526 | 5590156 | ||
BC101624.1 | - | 2 | 380 | 8069130 | ||
BC101626.1 | - | 2 | 380 | 8069132 |