Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301020.1 | NP_001287949.1 | 2 | 360 | ||
NM_004152.3 | NP_004143.1 | 2 | 366 | |||
GenBank mRNA | AF090094.1 | - | 1043 | |||
AK307221.1 | - | 2 | 332 | |||
BC010063.1 | - | 2 | 316 | |||
BI667301.1 | - | 2 | 337 | |||
BM423450.1 | - | 2 | 315 | |||
D78361.1 | - | 2 | 397 | |||
D87914.1 | - | 2 | 313 | |||
HY050868.1 | - | 2 | 366 | |||
U09202.1 | - | 2 | 298 |