Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004161.4 | NP_004152.1 | 4 | 599 | ||
GenBank mRNA | AF498929.1 | - | 4 | 213 | ||
AK315495.1 | - | 4 | 408 | |||
BC000905.2 | - | 4 | 391 | 2900705 | ||
BT019528.1 | - | 4 | 213 | |||
BT019529.1 | - | 4 | 213 | |||
BX571747.1 | - | 4 | 396 | |||
CR536488.1 | - | 4 | 213 | |||
M28209.1 | - | 4 | 263 | |||
X13898.1 | - | 4 | 213 |