Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008726.2 | NP_001008726.1 | 4 | 1770 | ||
NM_001258272.1 | NP_001245201.1 | 3 | 2166 | |||
NM_080666.3 | NP_542397.1 | 3 | 1661 | |||
XM_011536383.2 | XP_011534685.1 | 2 | 1700 | |||
XM_011536385.2 | XP_011534687.1 | 4 | 1668 | |||
GenBank mRNA | AF115513.1 | - | 3 | 1593 | ||
AK095324.1 | - | 3 | 1609 | |||
BC010698.1 | - | 2 | 1553 | 3870073 | ||
BX248008.1 | - | 3 | 1650 |