Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138396.5 | NP_612405.2 | 4 | 1464 | ||
GenBank mRNA | AK093897.1 | - | 2 | 379 | ||
BC009489.2 | - | 2 | 944 | 3936171 | ||
BC036455.2 | - | 4 | 1445 | 4811410 | ||
BC050397.1 | - | 4 | 1464 |