Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254755.1 | NP_001241684.1 | 4 | 1253 | ||
NM_001254756.1 | NP_001241685.1 | 4 | 1256 | |||
NM_144611.3 | NP_653212.1 | 4 | 1948 | |||
XM_006721447.2 | XP_006721510.2 | 4 | 3085 | |||
XM_011523649.2 | XP_011521951.1 | 4 | 1336 | |||
XM_017024180.1 | XP_016879669.1 | 4 | 1339 | |||
XM_017024181.1 | XP_016879670.1 | 4 | 1228 | |||
XM_017024182.1 | XP_016879671.1 | 4 | 3390 | |||
GenBank mRNA | AK172844.1 | - | 4 | 1414 | ||
BC051697.1 | - | 4 | 1860 | 5190840 |