Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008239.2 | NP_001008240.1 | 4 | 1400 | ||
NM_145055.3 | NP_659492.1 | 5 | 1583 | |||
XM_011525822.1 | XP_011524124.1 | 5 | 1434 | |||
XM_017025555.1 | XP_016881044.1 | 4 | 1247 | |||
GenBank mRNA | AK289738.1 | - | 4 | 1284 | ||
AK292981.1 | - | 4 | 1388 | |||
AL713661.1 | - | 3 | 580 | |||
AL832027.2 | - | 3 | 928 | |||
BC016149.1 | - | 4 | 1256 | 4040087 | ||
BC084579.1 | - | 4 | 1367 | 6141272 | ||
BX640683.1 | - | 3 | 1081 |