Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256442.1 | NP_001243371.1 | 3 | 1251 | ||
NM_001256443.1 | NP_001243372.1 | 2 | 1678 | |||
NM_145239.2 | NP_660282.2 | 3 | 1251 | |||
XM_011545715.2 | XP_011544017.1 | 3 | 1180 | |||
XM_011545716.2 | XP_011544018.1 | 3 | 1196 | |||
XM_017022887.1 | XP_016878376.1 | 3 | 1246 | |||
XM_017022888.1 | XP_016878377.1 | 3 | 1180 | |||
XM_017022889.1 | XP_016878378.1 | 3 | 1246 | |||
GenBank mRNA | AK074572.1 | - | 3 | 1145 | ||
AK092265.1 | - | 2 | 1523 | |||
AK098379.1 | - | 4 | 963 | |||
AK225785.1 | - | 3 | 1146 | |||
AK292393.1 | - | 3 | 1148 | |||
AL834185.1 | - | 3 | 1092 | |||
BC011405.1 | - | 3 | 1142 | 3868330 | ||
BC053594.1 | - | 3 | 1070 | 5729288 | ||
DA346262.1 | - | 1 | 547 |