Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152464.2 | NP_689677.1 | 2, 3 | 381 | ||
GenBank mRNA | AK223584.1 | - | 2, 3 | 315 | ||
AK291475.1 | - | 2, 3 | 304 | |||
AK311363.1 | - | 263 | ||||
AY074907.1 | - | 2, 3 | 291 | |||
BC033113.1 | - | 2, 3 | 298 | 5429060 | ||
DC327155.1 | - | 2, 3 | 381 |