Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324338.1 | NP_001311267.1 | 4 | 552 | ||
NM_152597.4 | NP_689810.3 | 4 | 649 | |||
XM_011521305.2 | XP_011519607.1 | 4 | 525 | |||
XM_011521306.1 | XP_011519608.1 | 4 | 938 | |||
XM_011521307.2 | XP_011519609.1 | 4 | 525 | |||
XM_011521309.2 | XP_011519611.1 | 4 | 525 | |||
GenBank mRNA | AK093308.1 | - | 4 | 631 | ||
AY260140.1 | - | 3 | 446 | |||
BC045191.2 | - | 4 | 641 | 5297527 |