Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152599.3 | NP_689812.3 | 1 | 402 | ||
GenBank mRNA | AK093092.1 | - | 1 | 355 | ||
AY129026.1 | - | 1 | 362 | |||
BC040487.1 | - | 1 | 321 | 5272039 |