Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM227B SIRNA Assays ›
Gene Symbol:
FAM227B
Gene Name:
family with sequence similarity 227 member B
Gene Aliases:
C15orf33
Chromosome Location:
Chr.15: 49326997 - 49620931 on Build GRCh38
UniGene ID:
Hs.712959
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
141534
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM227B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_152647.2
NP_689860.2
11
1234
XM_005254213.3
XP_005254270.1
11
1311
XM_005254214.3
XP_005254271.1
11
1244
XM_005254215.3
XP_005254272.1
10
1275
XM_005254216.3
XP_005254273.1
10
1209
XM_005254219.3
XP_005254276.1
11
1311
XM_005254220.3
XP_005254277.1
11
1311
XM_006720423.2
XP_006720486.1
12
1276
XM_006720424.2
XP_006720487.1
11
1311
XM_006720426.2
XP_006720489.1
9
1170
XM_006720427.3
XP_006720490.1
12
1282
XM_011521319.2
XP_011519621.1
11
3194
XM_011521320.1
XP_011519622.1
9
686
XM_011521321.2
XP_011519623.1
11
1311
XM_011521322.1
XP_011519624.1
8
650
XM_011521324.1
XP_011519626.1
11
1311
XM_011521325.2
XP_011519627.1
11
1311
XM_011521326.1
XP_011519628.1
10
1209
XM_017021990.1
XP_016877479.1
8
1068
XM_017021991.1
XP_016877480.1
12
1351
XM_017021992.1
XP_016877481.1
9
999
XM_017021993.1
XP_016877482.1
8
1068
XM_017021994.1
XP_016877483.1
7
969
XM_017021995.1
XP_016877484.1
11
1311
XM_017021996.1
XP_016877485.1
10
759
GenBank mRNA
AK057362.1
-
11
1232
BC048125.1
-
10
1134
5272014
BC048139.1
-
10
1134
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0