Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168215.1 | NP_001161687.1 | 6 | 546 | ||
NM_153229.2 | NP_694961.2 | 5 | 511 | |||
XM_006721730.3 | XP_006721793.1 | 5 | 469 | |||
GenBank mRNA | AK090637.1 | - | 5 | 466 | ||
AY358204.1 | - | 6 | 546 | |||
BC064507.1 | - | 5 | 439 | 6455254 | ||
DB072318.1 | - | 5 | 512 |