Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122890.2 | NP_001116362.1 | 4 | 2269 | ||
NM_001288702.1 | NP_001275631.1 | 4 | 2287 | |||
NM_001288703.1 | NP_001275632.1 | 4 | 1679 | |||
NM_001288704.1 | NP_001275633.1 | 3 | 1583 | |||
NM_153338.3 | NP_699169.2 | 3 | 2173 | |||
GenBank mRNA | AK074646.1 | - | 3 | 2159 | ||
BM720407.1 | - | 1 | 38 |