Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199777.1 | NP_001186706.1 | 4 | 816 | ||
NM_172240.2 | NP_758440.1 | 5 | 835 | |||
NR_037659.1 | 4 | 663 | ||||
NR_037660.1 | 3 | 644 | ||||
GenBank mRNA | AB053266.1 | - | 4 | 325 | ||
AK074772.1 | - | 5 | 573 | |||
AK123106.1 | - | 4 | 813 | |||
AK123625.1 | - | 3 | 283 | |||
AK295596.1 | - | 4 | 401 | |||
AL832918.1 | - | 3 | 300 | |||
BC026080.2 | - | 5 | 597 | 4827689 | ||
BU521431.1 | - | 3 | 313 | |||
EF011619.1 | - | 5 | 597 |