Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199777.1 | NP_001186706.1 | 11 | 1891 | ||
NM_172240.2 | NP_758440.1 | 12 | 1910 | |||
NR_037659.1 | 12 | 1761 | ||||
NR_037660.1 | 10 | 1719 | ||||
GenBank mRNA | AB053266.1 | - | 11 | 1400 | ||
AK027829.1 | - | 8 | 1109 | |||
AK074772.1 | - | 12 | 1648 | |||
AK123106.1 | - | 11 | 1888 | |||
AK123625.1 | - | 8 | 1108 | |||
AL832918.1 | - | 11 | 1397 | |||
BC026080.2 | - | 12 | 1672 | 4827689 | ||
BX648293.1 | - | 2 | 4512 | |||
EF011619.1 | - | 12 | 1672 |