Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173598.4 | NP_775869.3 | 4 | 597 | ||
XM_011538224.2 | XP_011536526.1 | 4 | 932 | |||
XM_011538225.2 | XP_011536527.1 | 4 | 1391 | |||
XM_011538226.2 | XP_011536528.1 | 4 | 932 | |||
XM_011538229.2 | XP_011536531.1 | 4 | 932 | |||
XM_017019208.1 | XP_016874697.1 | 4 | 932 | |||
XM_017019209.1 | XP_016874698.1 | 4 | 932 | |||
GenBank mRNA | AY345972.1 | - | 2 | 266 | ||
BC107106.1 | - | 2 | 261 | |||
BC107107.1 | - | 2 | 261 | |||
BC127603.1 | - | 2 | 266 | 40019272 |