Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178844.3 | NP_849172.2 | 5 | 737 | ||
NR_075083.2 | 5 | 798 | ||||
XM_017023027.1 | XP_016878516.1 | 5 | 767 | |||
XM_017023028.1 | XP_016878517.1 | 6 | 941 | |||
XM_017023029.1 | XP_016878518.1 | 7 | 1022 | |||
XM_017023030.1 | XP_016878519.1 | 7 | 1083 | |||
XM_017023031.1 | XP_016878520.1 | 5 | 828 | |||
XM_017023032.1 | XP_016878521.1 | 7 | 1784 | |||
XM_017023033.1 | XP_016878522.1 | 7 | 961 | |||
XM_017023034.1 | XP_016878523.1 | 6 | 880 | |||
XM_017023035.1 | XP_016878524.1 | 6 | 909 | |||
XM_017023036.1 | XP_016878525.1 | 5 | 1468 | |||
XM_017023039.1 | XP_016878528.1 | 5 | 767 | |||
GenBank mRNA | AK090431.1 | - | 5 | 794 | ||
AY601811.1 | - | 5 | 724 | |||
FJ889357.1 | - | 2 | 313 |