Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181783.3 | NP_861448.2 | 6 | 993 | ||
XM_011537980.2 | XP_011536282.1 | 5 | 768 | |||
GenBank mRNA | AJ697717.1 | - | 6 | 863 | ||
AK074973.1 | - | 5 | 620 | |||
AK290436.1 | - | 6 | 993 | |||
AK302755.1 | - | 6 | 993 | |||
AK310375.1 | - | 6 | 993 | |||
AK310393.1 | - | 6 | 993 | |||
BC117175.1 | - | 5 | 814 | 40125726 | ||
BC117177.1 | - | 5 | 815 | 40125728 | ||
BX538169.1 | - | 6 | 966 | |||
CR749244.1 | - | 6 | 945 | |||
CR749309.1 | - | 6 | 979 | |||
CR933632.1 | - | 6 | 944 |