Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181789.2 | NP_861454.2 | 5 | 835 | ||
XM_011521501.2 | XP_011519803.1 | 5 | 2692 | |||
XM_017022121.1 | XP_016877610.1 | 4 | 678 | |||
XM_017022122.1 | XP_016877611.1 | 5 | 866 | |||
XM_017022123.1 | XP_016877612.1 | 5 | 657 | |||
XM_017022124.1 | XP_016877613.1 | 5 | 709 | |||
XM_017022125.1 | XP_016877614.1 | 5 | 696 | |||
XM_017022126.1 | XP_016877615.1 | 5 | 709 | |||
GenBank mRNA | AK172756.1 | - | 5 | 671 | ||
AY358144.1 | - | 4 | 296 | |||
BC113397.1 | - | 5 | 321 | 8322449 | ||
BC113399.1 | - | 5 | 321 | 8322451 | ||
BX538105.1 | - | 5 | 344 |