Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304459.1 | NP_001291388.1 | 6 | 1406 | ||
NM_182608.3 | NP_872414.3 | 5 | 1317 | |||
GenBank mRNA | AF517119.1 | - | 1 | 100 | ||
AK125517.1 | - | 5 | 1366 | |||
BX647684.1 | - | 2 | 2617 | |||
BX649168.1 | - | 6 | 1152 |