Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA6L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291420.1 | NP_001278349.1 | 8 | 2925 | ||
NM_198181.3 | NP_937824.3 | 9 | 3057 | |||
XM_011521601.2 | XP_011519903.1 | 8 | 3608 | |||
XM_017022226.1 | XP_016877715.1 | 7 | 3044 | |||
GenBank mRNA | AK128268.1 | - | 5 | 2604 | ||
BC041854.1 | - | 8 | 2926 | 5270411 | ||
BF671239.1 | - | 1 | 75 | |||
CR936812.1 | - | 6 | 5460 |