Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA6L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291420.1 | NP_001278349.1 | 8 | 3093 | ||
NM_198181.3 | NP_937824.3 | 9 | 3225 | |||
XM_011521601.2 | XP_011519903.1 | 8 | 3776 | |||
XM_017022226.1 | XP_016877715.1 | 7 | 3212 | |||
GenBank mRNA | BC041854.1 | - | 8 | 3094 | 5270411 |