Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184720.1 | NP_001171649.1 | 5 | 734 | ||
NM_001184721.1 | NP_001171650.1 | 5 | 734 | |||
NM_004130.3 | NP_004121.2 | 5 | 734 | |||
XM_017006275.1 | XP_016861764.1 | 4 | 491 | |||
XM_017006276.1 | XP_016861765.1 | 3 | 1006 | |||
GenBank mRNA | AF087942.1 | - | 5 | 554 | ||
AK312283.1 | - | 5 | 570 | |||
BC000033.2 | - | 5 | 590 | 3504538 | ||
BC031096.2 | - | 4 | 495 | |||
CR536547.1 | - | 4 | 501 | |||
U31525.1 | - | 5 | 628 | |||
U44131.1 | - | 4 | 509 | |||
X79537.1 | - | 5 | 584 |