Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105528.1 | NP_001098998.1 | 11 | 1273 | ||
NM_001308126.1 | NP_001295055.1 | 11 | 1273 | |||
NM_198995.2 | NP_945346.2 | 12 | 1342 | |||
XM_011525948.1 | XP_011524250.1 | 12 | 1342 | |||
XM_011525951.1 | XP_011524253.1 | 12 | 1342 | |||
XM_011525954.2 | XP_011524256.1 | 12 | 1342 | |||
XM_011525955.1 | XP_011524257.1 | 12 | 1342 | |||
XM_017025721.1 | XP_016881210.1 | 12 | 1342 | |||
XM_017025722.1 | XP_016881211.1 | 12 | 1363 | |||
XM_017025723.1 | XP_016881212.1 | 12 | 1372 | |||
XM_017025724.1 | XP_016881213.1 | 6 | 906 | |||
XM_017025725.1 | XP_016881214.1 | 6 | 1523 | |||
GenBank mRNA | AK126038.1 | - | 12 | 1342 | ||
BC091517.1 | - | 12 | 1314 | 30389491 | ||
BC130535.1 | - | 11 | 1210 | 40146566 | ||
BC144178.1 | - | 11 | 1210 | 9052698 |