Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017.2 | NP_001008.1 | 3 | 145 | ||
GenBank mRNA | AK312060.1 | - | 3 | 139 | ||
BC000475.1 | - | 1 | 124 | 2961637 | ||
BC006772.1 | - | 1 | 126 | 2899987 | ||
BC029732.1 | - | 3 | 125 | 4388273 | ||
BC066322.1 | - | 3 | 139 | 4816284 | ||
BC100032.1 | - | 3 | 125 | 6025884 | ||
L01124.1 | - | 3 | 145 | |||
L05090.1 | - | 2 | 81 | |||
X79239.1 | - | 3 | 113 |