Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002687.3 | NP_002678.2 | 9 | 987 | ||
GenBank mRNA | AK223612.1 | - | 9 | 922 | ||
AK292579.1 | - | 9 | 971 | |||
AK300164.1 | - | 2 | 236 | |||
AK303136.1 | - | 5 | 578 | |||
AK304622.1 | - | 1 | 92 | |||
BC062602.1 | - | 9 | 898 | 5491466 | ||
U77718.1 | - | 9 | 917 | |||
Y09703.1 | - | 9 | 897 |