Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002754.4 | NP_002745.1 | 12 | 1199 | ||
NR_072996.1 | 10 | 1047 | ||||
GenBank mRNA | AB209586.1 | - | 9 | 2005 | ||
AF004709.1 | - | 12 | 1148 | |||
AF015256.1 | - | 12 | 1160 | |||
AF092535.1 | - | 12 | 1121 | |||
AF100546.1 | - | 12 | 1102 | |||
AK222922.1 | - | 10 | 926 | |||
BC000433.2 | - | 12 | 1141 | 2819932 | ||
BC001641.2 | - | 12 | 1141 | 2819932 | ||
BC004428.1 | - | 12 | 1141 | 2819932 | ||
BC085196.1 | - | 10 | 1017 | 6107307 | ||
U93232.1 | - | 12 | 1107 | |||
Y10488.1 | - | 12 | 1122 |