Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135651.2 | NP_001129123.1 | 6 | 788 | ||
NM_001135652.2 | NP_001129124.1 | 4 | 481 | |||
NM_002759.3 | NP_002750.1 | 6 | 1022 | |||
XM_011532987.2 | XP_011531289.1 | 6 | 722 | |||
XM_017004503.1 | XP_016859992.1 | 6 | 718 | |||
GenBank mRNA | AK290655.1 | - | 6 | 708 | ||
AK313818.1 | - | 5 | 545 | |||
AY302136.1 | - | 4 | 465 | |||
BC007769.2 | - | 5 | 564 | |||
BC022314.1 | - | 6 | 676 | |||
BC040851.1 | - | 6 | 788 | |||
BC057805.1 | - | 6 | 693 | |||
BC093676.1 | - | 4 | 493 | 7939521 | ||
BC101475.1 | - | 4 | 493 | 8068981 | ||
BC143228.1 | - | 4 | 492 | |||
BC143230.1 | - | 4 | 493 | 9051731 | ||
M35663.1 | - | 5 | 651 | |||
M85294.1 | - | 4 | 495 |