Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282232.1 | NP_001269161.1 | 7 | 1075 | ||
NM_001282233.1 | NP_001269162.1 | 6 | 932 | |||
NM_001282234.1 | NP_001269163.1 | 7 | 952 | |||
NM_002791.2 | NP_002782.1 | 7 | 1027 | |||
NR_104110.1 | 6 | 848 | ||||
XM_017021468.1 | XP_016876957.1 | 7 | 2363 | |||
GenBank mRNA | AK298920.1 | - | 6 | 900 | ||
AK302008.1 | - | 7 | 877 | |||
AK316223.1 | - | 7 | 941 | |||
BC002979.1 | - | 7 | 918 | 3542966 | ||
BC017882.1 | - | 7 | 940 | 4278372 | ||
BC022354.1 | - | 7 | 897 | 4292526 | ||
BC023659.1 | - | 7 | 945 | 4869452 | ||
BC070137.1 | - | 7 | 949 | 6458246 | ||
X59417.1 | - | 7 | 929 |