Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201538.1 | NP_001188467.1 | 7 | 984 | ||
NM_001201539.1 | NP_001188468.1 | 7 | 1069 | |||
NM_004042.4 | NP_004033.2 | 7 | 950 | |||
XM_011545522.1 | XP_011543824.1 | 7 | 936 | |||
GenBank mRNA | BC022389.1 | - | 7 | 1069 | 4609825 | |
BX107362.1 | - | 2 | 416 | |||
X97868.1 | - | 8 | 921 |