Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286076.1 | NP_001273005.1 | 6 | 932 | ||
NM_001297558.1 | NP_001284487.1 | 6 | 588 | |||
NM_002946.4 | NP_002937.1 | 6 | 781 | |||
GenBank mRNA | AK298759.1 | - | 6 | 541 | ||
AK300691.1 | - | 6 | 674 | |||
AK312956.1 | - | 6 | 545 | |||
BC001630.1 | - | 6 | 518 | 3538351 | ||
BC012157.1 | - | 6 | 570 | 4563651 | ||
BC021257.2 | - | 6 | 709 | 4111194 | ||
BX333932.2 | - | 6 | 510 | |||
CR450348.1 | - | 5 | 427 | |||
J05249.1 | - | 6 | 504 |