Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130420.1 | NP_001123892.1 | 30 | 3844 | ||
NM_003075.3 | NP_003066.2 | 28 | 4030 | |||
NM_139067.2 | NP_620706.1 | 29 | 3778 | |||
XM_005269101.1 | XP_005269158.1 | 29 | 4060 | |||
XM_005269102.1 | XP_005269159.1 | 29 | 4057 | |||
XM_005269103.1 | XP_005269160.1 | 28 | 3964 | |||
XM_005269104.1 | XP_005269161.1 | 30 | 3778 | |||
XM_011538693.2 | XP_011536995.1 | 28 | 3969 | |||
XM_017019884.1 | XP_016875373.1 | 29 | 3712 | |||
XM_017019885.1 | XP_016875374.1 | 29 | 3688 | |||
XM_017019886.1 | XP_016875375.1 | 28 | 3622 | |||
XM_017019887.1 | XP_016875376.1 | 27 | 3854 | |||
GenBank mRNA | AB209006.1 | - | 30 | 3844 | ||
AB209293.1 | - | 30 | 3750 | |||
AK025128.1 | - | 14 | 2203 | |||
AK293896.1 | - | 29 | 3665 | |||
BC009067.1 | - | 7 | 1678 | |||
BC013045.1 | - | 29 | 3704 | 4180388 | ||
BC026222.2 | - | 28 | 3949 | 4395012 | ||
CX165574.1 | - | 1 | 139 |