Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003090.3 | NP_003081.2 | 5 | 499 | ||
NR_135506.1 | 3 | 272 | ||||
NR_135507.1 | 2 | 225 | ||||
NR_135508.1 | 5 | 499 | ||||
GenBank mRNA | AJ130971.1 | - | 2 | 126 | ||
AJ130972.1 | - | 3 | 173 | |||
AK090986.1 | - | 2 | 833 | |||
AK223070.1 | - | 5 | 456 | |||
AK223110.1 | - | 5 | 421 | |||
AK290078.1 | - | 5 | 455 | |||
AK312200.1 | - | 5 | 430 | |||
AL538391.3 | - | 5 | 498 | |||
BC022816.1 | - | 5 | 452 | 4904205 | ||
BC067846.1 | - | 5 | 468 | 30330402 | ||
BC071717.1 | - | 1 | 407 | 4514975 | ||
X13482.1 | - | 5 | 451 |