Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODXL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018111.2 | NP_001018121.1 | 2 | 370 | ||
NM_005397.3 | NP_005388.2 | 2 | 370 | |||
GenBank mRNA | AK055816.1 | - | 3 | 346 | ||
AK223573.1 | - | 2 | 362 | |||
BC042466.1 | - | 2 | 322 | |||
BC093730.1 | - | 2 | 132 | 7939575 | ||
BC112035.1 | - | 2 | 144 | 8327503 | ||
BC143318.1 | - | 2 | 138 | 9051823 | ||
U97519.1 | - | 2 | 368 |