Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003923.2 | 10, 11 | 1592 | |||
GenBank mRNA | AF038499.2 | - | 10, 11 | 1441 | ||
AK296746.1 | - | 10, 11 | 1551 |