Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135255.1 | NP_001128727.1 | 7 | 961 | ||
NM_001135256.1 | NP_001128728.1 | 7 | 864 | |||
NM_005610.2 | NP_005601.1 | 7 | 964 | |||
GenBank mRNA | AK222779.1 | - | 7 | 867 | ||
AK299251.1 | - | 7 | 864 | |||
AK299410.1 | - | 5 | 575 | |||
AK312571.1 | - | 7 | 902 | |||
BC015123.1 | - | 5 | 625 | |||
BC053904.1 | - | 7 | 877 | 6526200 | ||
BC075836.1 | - | 7 | 832 | 6338602 | ||
X71810.1 | - | 7 | 878 | |||
X74262.1 | - | 7 | 889 |