Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143676.1 | NP_001137148.1 | 5 | 1086 | ||
NM_001143677.1 | NP_001137149.1 | 3 | 337 | |||
NM_001143678.1 | NP_001137150.1 | 3 | 516 | |||
NM_001291995.1 | NP_001278924.1 | 3 | 292 | |||
NM_005627.3 | NP_005618.2 | 3 | 292 | |||
XM_011536071.1 | XP_011534373.1 | 6 | 868 | |||
GenBank mRNA | AF153609.1 | - | 3 | 245 | ||
AK055077.1 | - | 5 | 1086 | |||
AK098509.1 | - | 3 | 260 | |||
AK293383.1 | - | 3 | 326 | |||
AK295422.1 | - | 3 | 260 | |||
AK298688.1 | - | 3 | 575 | |||
BC001263.1 | - | 3 | 245 | 3459056 | ||
BX649005.1 | - | 2 | 693 | |||
DA309338.1 | - | 3 | 370 | |||
EU518415.1 | - | 5 | 488 | |||
FM205707.1 | - | 3 | 280 | |||
FM205708.1 | - | 3 | 315 | |||
FM205709.1 | - | 5 | 935 | |||
FM205710.1 | - | 2 | 404 | |||
Y10032.1 | - | 3 | 245 |