Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270476.1 | NP_001257405.1 | 5 | 757 | ||
NM_006245.3 | NP_006236.1 | 5 | 740 | |||
NM_180976.2 | NP_851307.1 | 5 | 644 | |||
NM_180977.2 | NP_851308.1 | 3 | 422 | |||
GenBank mRNA | AB000634.1 | - | 5 | 575 | ||
AB000635.1 | - | 3 | 269 | |||
AB209861.1 | - | 3 | 319 | |||
AB451342.1 | - | 5 | 561 | |||
AB451357.1 | - | 5 | 561 | |||
AK122701.1 | - | 5 | 634 | |||
AK290604.1 | - | 5 | 553 | |||
AK291797.1 | - | 5 | 648 | |||
AK299696.1 | - | 5 | 613 | |||
BC001095.2 | - | 5 | 624 | 3507280 | ||
BC001175.1 | - | 5 | 551 | 3355395 | ||
BC010692.1 | - | 5 | 634 | 3853892 | ||
D78360.1 | - | 5 | 542 | |||
L76702.1 | - | 5 | 748 |