Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242413.2 | NP_001229342.1 | 4 | 445 | ||
NM_001282644.1 | NP_001269573.1 | 4 | 443 | |||
NM_001323265.1 | NP_001310194.1 | 4 | 500 | |||
NM_001323267.1 | NP_001310196.1 | 4 | 443 | |||
NM_006257.4 | NP_006248.1 | 4 | 445 | |||
XM_005252496.4 | XP_005252553.1 | 4 | 545 | |||
XM_005252497.4 | XP_005252554.1 | 4 | 547 | |||
XM_006717465.3 | XP_006717528.1 | 6 | 2940 | |||
XM_017016410.1 | XP_016871899.1 | 5 | 2851 | |||
GenBank mRNA | AK308099.1 | - | 4 | 418 | ||
AK312411.1 | - | 4 | 395 | |||
AY702977.1 | - | 3 | 345 | |||
BC101465.1 | - | 3 | 364 | 8068971 | ||
BC113359.1 | - | 3 | 364 | 8322411 | ||
BX647657.1 | - | 3 | 529 | |||
L01087.1 | - | 4 | 439 | |||
L07032.1 | - | 3 | 366 |