Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209277.1 | - | 14 | 2370 | ||
AK295868.1 | - | 5 | 626 | |||
AK302825.1 | - | 6 | 1139 | |||
AK309123.1 | - | 5 | 1054 | |||
BC117261.1 | - | 14 | 2306 | 40125812 | ||
BC143366.1 | - | 14 | 2306 | 9051871 | ||
BC143367.1 | - | 13 | 2207 | 9051872 | ||
L13858.1 | - | 14 | 2280 | |||
L20686.1 | - | 3 | 348 |