Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012425.3 | NP_036557.1 | 9 | 1413 | ||
NM_152724.2 | NP_689937.2 | 8 | 1301 | |||
XM_005252552.3 | XP_005252609.1 | 8 | 1363 | |||
GenBank mRNA | BC005993.1 | - | 9 | 1383 | 4050837 | |
BC008691.1 | - | 8 | 1572 | 3872071 | ||
BC015644.2 | - | 9 | 1373 | 4647836 | ||
BC107898.1 | - | 8 | 1280 | |||
BX647733.1 | - | 8 | 1277 |