Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP4K4 SIRNA Assays ›
Gene Symbol:
MAP4K4
Gene Name:
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases:
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location:
Chr.2: 101697703 - 101894690 on Build GRCh38
UniGene ID:
Hs.701013
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
1430
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP4K4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242559.1
NP_001229488.1
25
2971
NM_001242560.1
NP_001229489.1
26
3332
NM_004834.4
NP_004825.3
25
3098
NM_145686.3
NP_663719.2
26
3446
NM_145687.3
NP_663720.1
26
2890
XM_005264048.2
XP_005264105.1
28
3475
XM_005264050.3
XP_005264107.1
27
3313
XM_005264052.3
XP_005264109.1
27
3310
XM_005264057.3
XP_005264114.1
27
3244
XM_005264058.3
XP_005264115.1
26
3235
XM_005264059.3
XP_005264116.1
27
3151
XM_005264060.3
XP_005264117.1
26
3139
XM_005264062.3
XP_005264119.1
26
3082
XM_005264063.3
XP_005264120.1
26
3079
XM_005264064.3
XP_005264121.1
25
3073
XM_005264065.3
XP_005264122.1
26
2989
XM_005264066.3
XP_005264123.1
26
2986
XM_005264068.3
XP_005264125.1
25
2980
XM_005264069.2
XP_005264126.1
25
2950
XM_005264071.2
XP_005264128.1
24
2878
XM_006712865.2
XP_006712928.1
27
3220
XM_006712868.2
XP_006712931.1
26
2989
XM_006712869.2
XP_006712932.1
25
2890
XM_017005349.1
XP_016860838.1
28
3379
XM_017005350.1
XP_016860839.1
27
3313
XM_017005351.1
XP_016860840.1
26
3232
XM_017005352.1
XP_016860841.1
27
3244
XM_017005353.1
XP_016860842.1
27
3241
XM_017005354.1
XP_016860843.1
27
3220
XM_017005355.1
XP_016860844.1
26
3211
XM_017005356.1
XP_016860845.1
26
3142
XM_017005357.1
XP_016860846.1
27
3151
XM_017005358.1
XP_016860847.1
27
3148
XM_017005359.1
XP_016860848.1
26
3121
XM_017005360.1
XP_016860849.1
26
3118
XM_017005361.1
XP_016860850.1
26
3121
XM_017005362.1
XP_016860851.1
26
3118
XM_017005363.1
XP_016860852.1
26
3082
XM_017005364.1
XP_016860853.1
25
3070
XM_017005365.1
XP_016860854.1
26
3052
XM_017005366.1
XP_016860855.1
26
3028
XM_017005367.1
XP_016860856.1
26
3025
XM_017005368.1
XP_016860857.1
26
2956
XM_017005369.1
XP_016860858.1
25
2887
XM_017005370.1
XP_016860859.1
25
2887
XM_017005371.1
XP_016860860.1
24
2881
XM_017005372.1
XP_016860861.1
25
2797
XM_017005373.1
XP_016860862.1
24
2788
XM_017005374.1
XP_016860863.1
25
2797
XM_017005375.1
XP_016860864.1
24
2788
GenBank mRNA
AB013385.1
-
9
878
AB014587.1
-
24
2774
AF096300.1
-
25
2799
AK001272.1
-
9
1033
AK025610.1
-
13
1647
AK074592.1
-
5
639
AK295610.1
-
25
3116
AK296396.1
-
26
3028
AL137755.1
-
18
2427
AY212247.1
-
25
2966
AY834276.1
-
21
2375
EU794634.1
-
26
2940
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008696, NM_001252200, NM_001252201, NM_001252202, BC141109, and others.
Matching Taqman Gene Expression Assay:
Hs00377410_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction by protein phosphorylation
microvillus assembly
intracellular signal transduction
negative regulation of apoptotic process
regulation of JNK cascade
creatine kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0