Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021133.3 | NP_066956.1 | 2 | 449 | ||
GenBank mRNA | BC090934.1 | - | 2 | 433 | 3073811 | |
BC114433.1 | - | 1 | 315 | 40054016 | ||
BC115697.1 | - | 1 | 315 | 40054014 | ||
BU940316.1 | - | 2 | 449 | |||
CR627369.1 | - | 2 | 447 | |||
DA127960.1 | - | 2 | 449 |