Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021133.3 | NP_066956.1 | 7 | 3427 | ||
GenBank mRNA | AW968304.1 | - | 1 | 366 | ||
BC090934.1 | - | 8 | 3652 | 3073811 | ||
BX097443.1 | - | 1 | 91 |