Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323541.1 | NP_001310470.1 | 9 | 1177 | ||
NM_022039.3 | NP_071322.1 | 9 | 1727 | |||
NR_136613.1 | 8 | 1632 | ||||
XM_005270053.2 | XP_005270110.2 | 10 | 1850 | |||
GenBank mRNA | AF281859.1 | - | 9 | 2000 | ||
AK056917.1 | - | 5 | 2723 | |||
AK123173.1 | - | 3 | 2313 | |||
BC007380.2 | - | 7 | 1026 | |||
BC063415.1 | - | 9 | 1494 | 5223794 |