Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002786.3 | NP_002777.1 | 7 | 656 | ||
NM_148976.2 | NP_683877.1 | 8 | 873 | |||
GenBank mRNA | AK290765.1 | - | 7 | 597 | ||
BC002577.2 | - | 7 | 575 | 3140784 | ||
BC005932.1 | - | 7 | 591 | 4078894 | ||
BC008472.1 | - | 7 | 556 | 4282076 | ||
BC009576.1 | - | 7 | 564 | 4105838 | ||
BC015105.1 | - | 7 | 590 | 3950568 | ||
BC015356.1 | - | 7 | 559 | 3452219 | ||
BC022372.1 | - | 7 | 543 | 4770620 | ||
BT006647.1 | - | 6 | 510 | |||
D00759.1 | - | 7 | 551 | |||
FJ224300.1 | - | 8 | 855 | |||
M64992.1 | - | 8 | 855 | |||
X61969.1 | - | 7 | 615 |