Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142349.1 | NP_001135821.1 | 1, 2 | 409 | ||
NM_174934.3 | NP_777594.1 | 2, 3 | 472 | |||
NR_024527.1 | 1, 2 | 409 | ||||
GenBank mRNA | AK314531.1 | - | 2, 3 | 385 | ||
AY149967.1 | - | 2, 3 | 383 | |||
BC035017.2 | - | 1, 2 | 373 | 4817294 | ||
DA111034.1 | - | 1, 2 | 409 | |||
DA211092.1 | - | 1, 2 | 373 | |||
DA247019.1 | - | 1, 2 | 359 |